Order Entry
Poland
ContactUsLinkComponent
Rat Recombinant IGF-I
Rat Recombinant IGF-I
Catalog # APOSBITP1108-1MG
Supplier:  Apollo Scientific
CAS Number:  
Rat Recombinant IGF-I
Catalog # APOSBITP1108-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Protein synonyms:
    Insulin Like Growth Factor-I
  • Protein/peptide name:
    IGF-I

Specifications

About this item