0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Proteins and Peptides
Human recombinant FGF-a
Human recombinant FGF-a
APOSBITP1045-1MG
:
Apollo Scientific
:
Human recombinant FGF-a
APOSBITP1045-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Seq:
23
Protein synonyms:
Fibroblast Growth Factor-acidic
Protein/peptide name:
FGF-a