Order Entry
Poland
Orders LinkContactUsLinkComponent
Diacetoxyscirpenol Standard Solution

Diacetoxyscirpenol Standard Solution

Supplier:   Apollo Scientific
Diacetoxyscirpenol Standard Solution
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents

Analytical reference standard solution in acetonitrile.

Caution: Expires in 12 months.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • APOSBIMS125-1ML
  • APOSBIMS125-5ML
  • Description:
  • Diacetoxyscirpenol
  • Diacetoxyscirpenol
  • CAS No.:
  • 2270-40-8
  • 2270-40-8
  • Formula:
  • C₁₉H₂₆O₇
  • C₁₉H₂₆O₇
  • Flash point:
  • 2 °C
  • 2 °C
  • Storage temperature:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Concentration
    Pack type
    Pk
    Availability
    Price
  • APOSBIMS125-1ML
    Diacetoxyscirpenol
    Diacetoxyscirpenol
    100 µg/ml in acetonitrile
    Bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 1 240 zł
    1 240 zł
    1 * 1 mL
       1 240 zł

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIMS125-1ML
    Description
    Diacetoxyscirpenol
    CAS No.
    2270-40-8
    Formula
    C₁₉H₂₆O₇
    Flash point
    2 °C
    Storage temperature
    Freezer
  • APOSBIMS125-5ML
    Diacetoxyscirpenol
    Diacetoxyscirpenol
    100 µg/ml in acetonitrile
    Bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 4 850 zł
    4 850 zł
    1 * 5 mL
       4 850 zł

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIMS125-5ML
    Description
    Diacetoxyscirpenol
    CAS No.
    2270-40-8
    Formula
    C₁₉H₂₆O₇
    Flash point
    2 °C
    Storage temperature
    Freezer