Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Fusarenon X Standard Solution

Fusarenon X Standard Solution
 
Fusarenon X Standard Solution

 

 

Analytical reference standard solution in acetonitrile.

 : Expires in 12 months.

 

 

 

  • Catalog No:
  • APOSBIMS116-1ML
  • APOSBIMS116-5ML
  • Description:
  • Fusarenon X
  • Fusarenon X
  • CAS No.:
  • 23255-69-8
  • 23255-69-8
  • Formula:
  • C₁₇H₂₂O₈
  • C₁₇H₂₂O₈
  • Flash point:
  • 2 °C
  • 2 °C
  • Storage temperature:
  • Freezer
  • Freezer

 

Product Family Options

 

  • Description
    Concentration
    Pack type
    Pk
     
     
  • Fusarenon X
    100 µg/ml in acetonitrile
    Bottle
    1 mL
       
       
    1 * 1 mL
    1 * 1 mL - 955 zł
    955 zł
    1 * 1 mL
       955 zł

     
     
    Specifications
    Cat. No.
    APOSBIMS116-1ML
    Description
    Fusarenon X
    CAS No.
    23255-69-8
    Formula
    C₁₇H₂₂O₈
    Flash point
    2 °C
    Storage temperature
    Freezer
  • Fusarenon X
    100 µg/ml in acetonitrile
    Bottle
    5 mL
       
       
    1 * 5 mL
    1 * 5 mL - 3 270 zł
    3 270 zł
    1 * 5 mL
       3 270 zł

     
     
    Specifications
    Cat. No.
    APOSBIMS116-5ML
    Description
    Fusarenon X
    CAS No.
    23255-69-8
    Formula
    C₁₇H₂₂O₈
    Flash point
    2 °C
    Storage temperature
    Freezer