Order Entry
Poland
ContactUsLinkComponent
Deoxynivalenol and acetyl deoxynivalenol standard solutions
Deoxynivalenol and acetyl deoxynivalenol standard solutions
Supplier:  Apollo Scientific
Certificates
Deoxynivalenol and acetyl deoxynivalenol standard solutions
Supplier:  Apollo Scientific

Specifications

  • CAS No.:
    51481-10-8
  • Formula:
    C₁₅H₂₀O₆
  • Storage temperature:
    Freezer
  • Description:
    Deoxynivalenol (DON)
  • Flash point:
    2 °C

Specifications

Frequently Bought Together