

1-Ethynyl-3,5-difluorobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:151361-87-4
:
Formula:C₈H₄F₂
MDL Number:MFCD04039982
Boiling Pt:123…124 °C
Density:1.163 g/cm³ (25 °C)
Flash Pt:33 °C
UN:1993
ADR:3,III
:138.12 g/mol
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together