
1H,1H-Tricosafluoro-1-dodecanol 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:423-65-4
:
Formula:C₁₂H₃F₂₃O
MDL Number:MFCD00153235
:600.12 g/mol
Boiling Pt:133 °C/30mmHg
Melting Pt:111…113 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together