
3,4-Difluorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113028-75-4
:
Formula:C₇H₃F₂NS
MDL Number:MFCD00175109
:171.17 g/mol
Boiling Pt:170 °C
Density:1.35 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together