Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-tert-Butyl-2-nitro-4-(trifluoromethyl)aniline
N-tert-Butyl-2-nitro-4-(trifluoromethyl)aniline
N-tert-Butyl-2-nitro-4-(trifluoromethyl)aniline
APOSPC9341-1G
:
Apollo Scientific
:
N-tert-Butyl-2-nitro-4-(trifluoromethyl)aniline
APOSPC9341-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
:
167…168 °C
:
GHS07
:
Refrigerator
:
247113-96-8
:
C₁₁H₁₃F₃N₂O₂
:
MFCD00707485