Order Entry
Poland
ContactUsLinkComponent
3,5-Dibromo-2-fluoro-4-methylpyridine 97%
3,5-Dibromo-2-fluoro-4-methylpyridine 97%
Catalog # APOSPC9264-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dibromo-2-fluoro-4-methylpyridine 97%
Catalog # APOSPC9264-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.9100 g/mol
Storage Temperature:Ambient
CAS Number:1000340-01-1
Formula:C₆H₄NBr₂F
MDL Number:MFCD09864703