Order Entry
Poland
ContactUsLinkComponent
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate
Irritant
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate
Catalog # APOSPC8912-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2-(trifluoromethyl)pyrimidine-4-carboxylate
Catalog # APOSPC8912-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.12 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:878745-51-8
Formula:C₇H₅F₃N₂O₂
MDL Number:MFCD05662690