Order Entry
Poland
ContactUsLinkComponent
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Irritant
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Catalog # APOSPC8786-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-{5-Methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}acetic acid
Catalog # APOSPC8786-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :301.29 g/mol
Melting Pt:119…121 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:924868-87-1
Formula:C₁₃H₁₀F₃NO₂S
MDL Number:MFCD08443969