Methyl-4'-chloro-3'-fluoro[1,1'-biphenyl]-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886362-12-5
:
Formula:C₁₄H₁₀ClFO₂
MDL Number:MFCD06801758
:264.69 g/mol
Melting Pt:88…90 °C
Storage Temperature:Ambient
Frequently Bought Together