Methyl-5-iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridinecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:937602-50-1
:
Formula:C₁₅H₁₁F₃INO₃
MDL Number:MFCD09027149
:437.16 g/mol
Melting Pt:134…136 °C
Storage Temperature:Ambient
Frequently Bought Together