2-(4-Bromo-2-fluorophenyl)-1,3-thiazolane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:937602-46-5
:
Formula:C₉H₉BrFNS
MDL Number:MFCD09027142
:262.15 g/mol
Melting Pt:70…72 °C
Storage Temperature:Ambient
Frequently Bought Together