
2-[Cyano(3,4-dimethoxyphenyl)methyl]-6-fluorobenzenecarbonitrile 97%
Catalog # APOSPC8439-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:296.3 g/mol
Melting Pt:160…161 °C
:GHS07
Storage Temperature:Ambient
CAS Number:339012-94-1
Formula:C₁₇H₁₃FN₂O₂
MDL Number:MFCD00231998