3-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzenesulphonamide
Catalog # APOSPC8109-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:97…99 °C
Storage Temperature:Refrigerator
CAS Number:951884-65-4
Formula:C₉H₉BrF₃NO₂S
MDL Number:MFCD09801011