3-Bromo-N-(furan-2-ylmethyl)-5-(trifluoromethyl)benzenesulphonamide
Catalog # APOSPC8105-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:951884-84-7
Formula:C₁₂H₉BrF₃NO₃S
MDL Number:MFCD09801006