Order Entry
Poland
ContactUsLinkComponent
N-(4-Fluorophenyl) 4-boronobenzenesulphonamide
N-(4-Fluorophenyl) 4-boronobenzenesulphonamide
Catalog # APOSPC8049-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Fluorophenyl) 4-boronobenzenesulphonamide
Catalog # APOSPC8049-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :295.09 g/mol
Storage Temperature:Ambient
CAS Number:957121-13-0
Formula:C₁₂H₁₁BFNO₄S
MDL Number:MFCD09800889