
1-(3,4-Difluorophenyl)biguanide hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:537037-56-2
:
Formula:C₈H₁₀ClF₂N₅
MDL Number:MFCD03094449
Melting Pt:213…215 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together