
1-(3,5-Difluorophenyl)biguanide hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1030829-66-3
:
Formula:C₈H₁₀ClF₂N₅
MDL Number:MFCD03094447
Melting Pt:226 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together