
Ethyl-2,3,4,5-tetrafluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:122894-73-9
:
Formula:C₉H₆F₄O₂
MDL Number:MFCD02094158
:222.14 g/mol
Boiling Pt:238…239 °C
Density:1.382 g/cm³ (25 °C)
Flash Pt:95 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together