
2-Difluoromethyl-1H-benzoimidazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:705-09-9
:
Formula:C₈H₆F₂N₂
MDL Number:MFCD00463015
:168.15 g/mol
Melting Pt:155…157 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together