
1H,1H-Nonafluoro-3,6-dioxaheptan-1-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:330562-43-1
:
Formula:C₅H₃F₉O₃
MDL Number:MFCD03094428
:282.06 g/mol
Boiling Pt:117 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together