
4-(Trifluoromethoxy)phenylhydrazine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:133115-72-7
:
Formula:C₇H₈ClF₃N₂O
MDL Number:MFCD00053033
:228.60 g/mol
Melting Pt:230 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together