Order Entry
Poland
ContactUsLinkComponent
8-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-dioxa-8-azaspiro[4.5]decane
8-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-dioxa-8-azaspiro[4.5]decane
Catalog # APOSPC7334-1G
Supplier:  Apollo Scientific
CAS Number:  
8-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-dioxa-8-azaspiro[4.5]decane
Catalog # APOSPC7334-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :288.27 g/mol
Melting Pt:95…96 °C
Storage Temperature:Ambient
CAS Number:942474-71-7
Formula:C₁₃H₁₅F₃N₂O₂
MDL Number:MFCD08692463