
2,3,6-Trifluorobenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2358-29-4
:
Formula:C₇H₃F₃O₂
MDL Number:MFCD00002409
:176.09 g/mol
Melting Pt:130…131 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together