


Boron trifluoride dihydrate 96%
:13319-75-0
:
:BF₃·2H₂O
:MFCD00149901
:103.84 g/mol
:58…60 °C/1.5mmHg
:6 °C
:1.636 g/cm³ (25 °C)
:>100 °C
:2851
:8,II
:GHS05
:GHS06
:GHS08
:Ambient
Frequently Bought Together
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals