

1H,1H-Heptacosafluoro-1-tetradecanol 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15622-57-8
:
Formula:C₁₄H₃F₂₇O
MDL Number:MFCD00153264
:700.13 g/mol
Boiling Pt:211 °C/100mmHg
Melting Pt:125…126 °C
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,III
:GHS05
:GHS06
Frequently Bought Together