
1H,1H,2H,2H-Perfluorooctyl methacrylate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2144-53-8
:
Formula:C₁₂H₉F₁₃O₂
MDL Number:MFCD00077580
:432.18 g/mol
Boiling Pt:92 °C/8mmHg
Melting Pt:–33 °C
Density:1.496 g/cm³ (25 °C)
Flash Pt:113 °C
EINECS:218-407-9
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together