

1H,1H,2H-Perfluoro-1-octene
Catalog # APOSPC6163-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:346.09 g/mol
Boiling Pt:106…107 °C
Density:1.58 g/cm³ (25 °C)
Flash Pt:20 °C
Storage Temperature:Refrigerator
EINECS:246-791-8
UN:1993
ADR:3,II
:GHS02
:GHS07
CAS Number:25291-17-2
Formula:C₈H₃F₁₃
MDL Number:MFCD00039249