
1H,1H-Nonadecafluoro-1-decanol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:307-37-9
:
Formula:C₁₀H₃F₁₉O
MDL Number:MFCD00153234
:500.1 g/mol
Boiling Pt:95 °C/20mmHg
Melting Pt:83…90 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together