
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306-94-5
:
Formula:C₁₀F₁₈
MDL Number:MFCD00010626
:462.08
Boiling Pt:141…143 °C
Melting Pt:–10 °C
Density:1.908 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:206-192-4
:GHS07
Frequently Bought Together