6-Chloro-3,5-dibromo-2,2,3,4,4,5,6,6-octafluorohexanenitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:240800-52-6
:
MDL Number:MFCD08461581
:433.32 g/mol
Storage Temperature:Temperatura otoczenia
Frequently Bought Together