
2-Amino-5,6-difluorobenzoic acid
Catalog # APOSPC52215-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:173.12 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:442134-72-7
Formula:C₇H₅F₂NO₂
MDL Number:MFCD06808540