Order Entry
Poland
ContactUsLinkComponent
1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene 95%
1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene 95%
Catalog # APOSPC520648-5G
Supplier:  Apollo Scientific
CAS Number:  
1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene 95%
Catalog # APOSPC520648-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :212.58 g/mol
Storage Temperature:Ambient
CAS Number:1138444-89-9
Formula:C₈H₅ClF₄
MDL Number:MFCD11226702