
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:39637-99-5
:
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD00044400
:252.62 g/mol
Boiling Pt:213…214 °C
Density:1.35 g/cm³ (25 °C)
Flash Pt:89 °C
Storage Temperature:Freezer
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together