
Methyl-2-(bromomethyl)-5-fluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138786-65-9
:
Formula:C₉H₈BrFO₂
MDL Number:MFCD04114323
:247.06 g/mol
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together