2-{1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-indol-3-yl}acetohydrazide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339099-27-3
:
Formula:C₁₆H₁₂ClF₃N₄O
MDL Number:MFCD00664547
:368.75 g/mol
Melting Pt:151…153 °C
Storage Temperature:Ambient
Frequently Bought Together