
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18931-60-7
:
Formula:C₁₀H₆ClF₃O₂
MDL Number:MFCD00511276
:250.61 g/mol
Melting Pt:60…64 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together