
4-Fluoro-2-methoxy-5-nitroaniline 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1075705-01-9
:
Formula:C₇H₇FN₂O₃
MDL Number:MFCD23098941
:186.14 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together