

2,3-Difluoro-4-(difluoromethyl)pyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1374659-38-7
:
Formula:C₆H₃F₄N
MDL Number:MFCD22199287
:165.09 g/mol
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together