
2-Chloro-3-(difluoromethyl)pyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:865663-96-3
:
Formula:C₆H₄ClF₂N
MDL Number:MFCD16140197
:163.56 g/mol
Density:1.388 g/cm³ (25 °C)
Flash Pt:80.56 °C
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
:GHS06
Frequently Bought Together