Order Entry
Poland
ContactUsLinkComponent
Ethyl 4,6-dioxo-5[3-(trifluoromethyl)phenyl]-1,3A,4,5,6,6A-hexahydropyrrolo[3,4-C]pyrazole-3-carboxylate
Ethyl 4,6-dioxo-5[3-(trifluoromethyl)phenyl]-1,3A,4,5,6,6A-hexahydropyrrolo[3,4-C]pyrazole-3-carboxylate
Catalog # APOSPC4886-500MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4,6-dioxo-5[3-(trifluoromethyl)phenyl]-1,3A,4,5,6,6A-hexahydropyrrolo[3,4-C]pyrazole-3-carboxylate
Catalog # APOSPC4886-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
Melting Pt:172…174 °C
 :355.27 g/mol
Storage Temperature:Ambient
CAS Number:321522-08-1
Formula:C₁₅H₁₂F₃N₃O₄
MDL Number:MFCD01315600