

α,α,α,2,3,4,5-Heptafluorotoluene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:654-53-5
:
Formula:C₇HF₇
MDL Number:MFCD01862013
:218.07 g/mol
Boiling Pt:100…106 °C
Flash Pt:22 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together