
4-(4-Fluorophenyl)-3-thiosemicarbazide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:330-94-9
:
Formula:C₇H₈FN₃S
MDL Number:MFCD00041294
:185.22 g/mol
Melting Pt:171…172 °C (dec.)
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together