

1H,1H,2H-Perfluoro-1-pentene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:355-08-8
:
Formula:C₅H₃F₇
:196.07 g/mol
Boiling Pt:30…31 °C
Density:1.3391 g/cm³ (25 °C)
Flash Pt:100 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together