
(S)-1-(2,6-Dichloro-3-fluorophenyl)ethanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:877397-65-4
:
Formula:C₈H₇Cl₂FO
MDL Number:MFCD09863793
:209.04 g/mol
:GHS07
Storage Temperature:Temperatura otoczenia
Frequently Bought Together