Order Entry
Poland
ContactUsLinkComponent
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Corrosive
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Catalog # APOSPC448000-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Catalog # APOSPC448000-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :240.05 g/mol
Melting Pt:34 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
 :GHS05
CAS Number:402846-78-0
Formula:C₉H₁₇BF₄N₂
MDL Number:MFCD03427618