Order Entry
Poland
ContactUsLinkComponent
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester
Catalog # APOSPC430149-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester
Catalog # APOSPC430149-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :308.4 g/mol
Storage Temperature:Ambient
CAS Number:907572-24-1
Formula:C₁₇H₂₅FN₂O₂
MDL Number:MFCD22421729